Natriumhydroksid 95.0-100.5%, kuler, GenAR® NF, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

7772-06EA 2175 NOK
7772-06 7772-30 7772-88 7772-25 7772-26 7772-28
Natriumhydroksid 95.0-100.5%, kuler, GenAR® NF, Macron Fine Chemicals™
Natriumhydroksid
Formel: NaOH
Kokepunkt: 1390 °C (1013 hPa)
Smeltepunkt: 323 °C
Tetthet: 2,13 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Indeks: 14,08627

Bestill nå

Spesifikasjon Testresultater

Meets F.C.C. Requirements
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Food GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical 95.0 - 100.5 %
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Insoluble Substances and Organic Matter Passes Test
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Potassium (K) ≤0.5 %
NF/FCC - Carbonate (as Na₂CO₃) ≤3.0 %
FCC - Assay (Total alkali as NaOH) 95.0 - 100.5 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg)(mg/kg) ≤0.1
FCC - Arsenic (As)(mg/kg) ≤3
FCC - Identification Passes Test
FCC - Insoluble Substances and Organic Matter Passes Test
BP/EP - Assay (Total alkali as NaOH) 97.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤2.0 %
JP - Mercury (Hg) Passes Test
Endotoxin Concentration (EU/g) ≤5
Appearance (white hygroscopic pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR