E-64
Leverandør: G Bioscienes
786-049
786-049EA 8425 NOK
786-049
E-64
Proteiner
E-64 is an irreversible inhibitor of cysteine proteases. It interacts with the Sn subsites of proteases but has no action on cysteine residues in other proteins.