Cleaning solutions/spray for electrophoresis gel plates, GELPLATE-clean™
Leverandør: G Bioscienes
786-140 786-140RF 786-140RF-II 786-140RF-I
786-140EA 1775 NOK
786-140 786-140RF 786-140RF-II 786-140RF-I
Cleaning solutions/spray for electrophoresis gel plates, GELPLATE-clean™
Rengjøringsløsninger Dekontaminanter
GELPLATE-clean™ removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. It is suitable for sequencing and protein electrophoresis gel plates.
  • Prevent poor electrophoretic band migration
  • Prevent band distortions
  • Prevent poor image development
Leveringsinformasjon: Supplied as 2 solutions in 2× 250 ml spray bottles. Refill solutions are available and are offered separately.