Nucleic acid and protein destain, Silver D-Stain™
Leverandør: G Bioscienes
786-199
786-199EA 6175 NOK
786-199
Nucleic acid and protein destain, Silver D-Stain™
Elektroforese reagenser Reagenser for fjerning av fargeløsning
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.
Suitable for reducing the staining of silver stained gels and revealing over-stained bands.