Apoptotic DNA ladder kit
Leverandør: G Bioscienes
786-209
786-209EA 8625 NOK
786-209
Apoptotic DNA ladder kit
Elektroforese reagenser Molekylvekt markører
The Apoptotic DNA Ladder Kit contains the reagents necessary for the preparation of cellular fractions for the production of apoptosis nucleosomal DNA ladders. The kit does not require toxic phenol and DNA ladder is prepared either with or without genomic DNA.