Protein Assay, Protn-Latex™
Leverandør: G Bioscienes
786-20LATEX
786-20LATEXEA 15050 NOK
786-20LATEX
Protein Assay, Protn-Latex™
Assay Proteinanalyser
A simple and reliable method for estimation of protein contamination in latex. Detects as low as 25 to 40 μg protein/gm latex (or 25 to 50 ppm). Protein solutions applied to the test strips form compact and symmetrical spots, their diameter being directly proportional to protein concentration. Protn-Latex™ latex protein extraction buffer extracts protein from the latex sample, which is then applied to the test strips and rapidly developed. By measuring the Ø of protein spots with the Protn-Latex™ card, the range of protein concentration in the sample can be determined.
Bestillingsinformasjon: Available with or without an optional applicator device.