Vann, Proteomics Grade

Leverandør: G Bioscienes

Synonymer: Hydrogen oxide

786-229
786-229EA 1150 NOK
786-229
Vann, Proteomics Grade
Vann

Ultra pure proteomic grade water that is fully compatible with 2D electrophoresis and mass spectrometry analysis.

Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR