Proteome kits, FOCUS™ Bacterial Proteome
Leverandør: G Bioscienes
786-258
786-258EA 11150 NOK
786-258
Proteome kits, FOCUS™ Bacterial Proteome
Proteinekstraksjons Protein Extraction Kits
Specifically designed for bacterial research and supplied with bacteria specific reagents. Extracts and solubilises nearly all of the proteins from E.coli, including membrane as well as soluble proteins. Extraction is based on the gentle lysis of bacterial cells with LongLife™ Lysozyme enzyme, followed by extraction of total proteome with the supplied strong chaotropic extraction buffer that solubilises even the most difficult proteins.
  • Simple extraction protocol
  • Supplied with bacterial specific lytic enzyme preparation
  • Supplied with a strong proprietary chaotropic extraction buffer
  • Suitable for extraction from 50×50 μl bacterial cell pellet
  • Suitable for sample preparation for 2D gel electrophoresis and other applications