Recombinant protein purification resins, IMAC
Leverandør: G Bioscienes
786-542 786-543 786-285 786-286 786-287 786-403 786-402 786-455 786-454 786-456
786-542EA 2650 NOK
786-542 786-543 786-285 786-286 786-287 786-403 786-402 786-455 786-454 786-456
Recombinant protein purification resins, IMAC
Proteinrense Protein Purification Resins
Bestill nå
SPESIFIKASJONER
Beskrivelse His binding/wash buffer His elution buffer Copper chelating resin, for the purification of 6X His proteins Cobalt chelating resin, for the purification of 6X His proteins Zinc chelating resin, for the purification of 6X His proteins Cobalt chelating resin, 1 ml columns, for the purification of 6X His proteins Cobalt chelating resin, 0,2 ml columns, for the purification of 6X His proteins Cobalt chelating resin, 3 ml columns, for the purification of 6X His proteins
Environmentally Preferable
Liganddensitet - - 20 - 40 μmoles Cu2+/ml 20 - 40 μmoles Cu2+/ml 20 - 40 μmoles Zn2+/ml 20 - 40 μmoles Cu2+/ml 20 - 40 μmoles Cu2+/ml 20 - 40 μmoles Cu2+/ml
Bindingskapasitet - - >50 mg/ml >50 mg/ml >50 mg/ml >50 mg/ml >50 mg/ml >50 mg/ml