BLOT-FastStain™
Leverandør: G Bioscienes
786-34
786-34EA 7275 NOK
786-34
BLOT-FastStain™
Farger Elektroforese Farging
BLOT-FastStain™ is a unique stain for reversible staining of protein on nitrocellulose and PVDF transfer membranes. It only stains protein and leaves the background absolutely untouched and brilliant white resulting in high band visibility.
  • Rapid and sensitive staining of proteins on western blots
  • Reversible stain for protein membranes
  • Compatible with PVDF and nitrocellulose membranes
  • Detect >2 ng protein
BLOT-FastStain™ only stains protein and leaves the background absolutely untouched and brilliant white resulting in high band visibility. The staining procedure takes 10 minutes and has a sensitivity of 2ng BSA, higher than silver stains. Destain the membrane by simply rinsing in warm water for 10 minutes.
Staining and destaining does not affect the biological properties of the proteins. After destaining, protein bands can be probed with Western blot protocols and other analyses including sequencing work.
BLOT-FastStain™ is fully compatable with the LiCor Odyssey detection system and has succesfully been used as a loading control in this Odyssey system.
Application: Staining of membranes for visualization of proteins onto the membrane after Western transfer and dot-blot applications.
NB: Do not store in the cold.