Proteome kits, FOCUS™ Insect Proteome
Leverandør: G Bioscienes
786-360
786-360EA 8750 NOK
786-360
Proteome kits, FOCUS™ Insect Proteome
Proteinekstraksjons Protein Extraction Kits
Extracts and solubilises nearly all of the proteins from insect cell cultures (i.e. Sf9 and Sf21), including membrane as well as soluble proteins, using a strong chaotropic extraction buffer to solubilise even the most difficult proteins.
  • Single step extraction protocol
  • Supplied with a strong proprietary chaotropic extraction buffer
  • For the extraction of protein from 50×50 μl insect cell pellets
  • Suitable for sample preparation for 2D gel electrophoresis and other applications