Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Leverandør: G Bioscienes
786-499
786-499EA 2050 NOK
786-499
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Elektroforese reagenser Reagenser for fjerning av fargeløsning
After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution.
  • Contains Methanol and Acetic acid