Immobilised avidin for biotin purification
Leverandør: G Bioscienes
786-594 786-593
786-594EA 24200 NOK
786-594 786-593
Immobilised avidin for biotin purification
Proteinrense Protein Purification Resins
This avidin resin is designed for the single step small and large scale affinity purification of proteins and antibodies with a biotin tag. The resin can also be used for immunoprecipitations using biotin labelled antibodies.