Film cleaner, Swift™
Leverandør: G-Biosciences
786-678
786-678EA 4600 NOK
786-678
Film cleaner, Swift™
Filmbehandlingsutstyr
Swift™ Film Cleaner allows researchers to clean film that has been overexposed or has a high background or speckling without having to repeat experiments. Suitable for all exposed film developed for gel shift assays, Western, Southern and Northern blots.
  • Save time by eliminating need to repeating experiment
  • Provides even signal removal to preserve results
  • Rapidly stopped once optimal signal achieved