Destaining solution, Blue-OUT™ Mass spectrometry
Leverandør: G Bioscienes
Danger
786-683
786-683EA 3400 NOK
786-683
Destaining solution, Blue-OUT™ Mass spectrometry
Destaining solution
Blue-OUT™ for the removal of Coomassie and fluorescent stain prior to mass spectrometry
Blue-OUT™ is a unique product that washes the interfering Coomassie and fluorescent protein stains from gel pieces prior to their treatment for protein sequencing or mass spectrometry.
Blue-OUT™ is also supplied in the InGel™ Blue kit for the tryptic digestion and peptide extraction of Coomassie stained proteins.
Lagringstemperatur: Romtemperatur
FN: 1648
ADR: 3 II