Protein solubilisation buffer
Leverandør: G Bioscienes
786-PSB
786-PSBEA 4975 NOK
786-PSB
Protein solubilisation buffer
Buffere Buffere, ekstrasjons
FOCUS™ protein solubilisation buffer is a dry, urea-based buffer for protein solubilisation. It is a pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulphobetaine for maximum solubilising strength.
  • Convenient and simple to use - simply hydrate and use
  • Prevents urea induced protein carbamylation
  • Prevents waste of unused reagents
Leveringsinformasjon: Supplied with specific dilution buffer for optimal rehydration and protein solubilisation conditions.