Rhodamin B, Supelco®

Leverandør: Merck
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

M-Clarity™ quality level = MQ100

79754-25MG
79754-25MGEA 1625 NOK
79754-25MG
Rhodamin B, Supelco®
Rhodamin B
Formel: C₂₈H₃₁ClN₂O₃
Smeltepunkt: 199…201 °C
Tetthet: 1,31 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
EINECS: 201-383-9
Merck Indeks: 13,08266
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d4f879055b2eedc029b9e73fa00ac2cfe217f5e&vid=d8f6a98242a125c19372c39843173e429ad1b89a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR