Supelco® Organic Reference Standard, Azodicarboxamide

Leverandør: Merck

79997-50MG
79997-50MGEA 725 NOK
79997-50MG
Supelco® Organic Reference Standard, Azodicarboxamide
Standarder Organiske standarder

  • GHS signal: GHS08


NB: P210 - P240 - P241 - P261 - P280 - P304 + P340 + P312

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80fb66c758aae13f1c13eed3e7bb8cbbacac2544&vid=80fb66c758aae13f1c13eed3e7bb8cbbacac2544&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 123-77-3
MDL No. MFCD00007958
Formel NH₂CON=NCONH₂
Molekylvekt 116,08
Smeltepunkt 220 - 225 °C (dec.) (lit.)
Hardhetsklasse beskrivelse H228 - H334
Flammepunkt >200 °C
Fraktforhold STD
Lagringsforhold RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR