Acetamidine hydrochloride

Leverandør: Merck

8.00026.9020
8.00026.9020EA 0 NOK
8.00026.9020
Acetamidine hydrochloride
Acetamidine hydrochloride
Formel: C₂H₇ClN₂
MW: 94.54 g/mol
MDL nummer: MFCD00013016
CAS-nummer: 124-42-5
EINECS: 204-700-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR