N-(4-aminophenyl)aniline hydrochloride for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 4-Aminodiphenylamine hydrochloride

M-Clarity™ quality level = MQ200

8.00405.0100
8.00405.0100EA 1200 NOK
8.00405.0100
N-(4-aminophenyl)aniline hydrochloride for synthesis, Sigma-Aldrich®
N-(4-aminophenyl)aniline hydrochloride
Formel: C₁₂H₁₃ClN₂
Lagringstemperatur: Romtemperatur
CAS-nummer: 56426-15-4
EINECS: 260-173-5

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) slightly soluble
Melting Point 258 - 261 °C
Molar Mass 220.7 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR