Jodetan stabilised for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Ethyl iodide

Stabilisering: Stabilised with silver
M-Clarity™ quality level = MQ200

8.00896.0100 8.00896.0500 8.00896.9050
8.00896.0100EA 1530 NOK
8.00896.0100 8.00896.0500 8.00896.9050
Jodetan stabilised for synthesis, Sigma-Aldrich®
Jodetan
Formel: C₂H₅I
Kokepunkt: 72 °C (1013 hPa)
Smeltepunkt: –108 °C
Tetthet: 1,936 g/cm³ (20 °C)
Flash punkt: 53 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001091
CAS-nummer: 75-03-6
EINECS: 200-833-1
FN: 1993
ADR: 3,III
Merck Indeks: 13,03846

Bestill nå

Spesifikasjon Testresultater

Solubility 4 g/l (20 °C) (slow decomposition)
Melting Point -108 °C
Molar Mass 155.97 g/mol
Boiling Point 69 - 73 °C (1013 hPa)
Vapor Pressure 133 hPa (18 °C)
Flash Point 53 °C
Refractive Index 1.513 (20 °C, 589 nm)
Density 1.93 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR