2-Aminobenzothiazole, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II

M-Clarity™ quality level = MQ200

8.01268.0050 8.01268.0250
8.01268.0050EA 363 NOK
8.01268.0050 8.01268.0250
2-Aminobenzothiazole, Sigma-Aldrich®
2-Aminobenzothiazole
Formel: C₇H₆N₂S
Kokepunkt: 190…195 °C (0,05 mmHg)
Smeltepunkt: 127…130 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005785
CAS-nummer: 136-95-8
EINECS: 205-268-4
Merck Indeks: 13,00423

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 127 - 130 °C
Molar Mass 150.2 g/mol
Synonyms 2-Benzothiazolylamine
Hill Formula C7H6N2S


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR