2-klor-2-metylpropan for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.01550.0100 8.01550.1000 8.01550.9160
8.01550.0100EA 375 NOK
8.01550.0100 8.01550.1000 8.01550.9160
2-klor-2-metylpropan for syntese, Sigma-Aldrich®
2-klor-2-metylpropan
Formel: C₄H₉Cl
Kokepunkt: 51 °C (1013 hPa)
Smeltepunkt: –28 °C
Tetthet: 0,85 g/cm³ (20 °C)
Flash punkt: –27 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000816
CAS-nummer: 507-20-0
EINECS: 208-066-4
FN: 1127
ADR: 3,II
Merck Indeks: 13,01561

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble,(slow decomposition)
Melting Point -28 °C
Molar Mass 92.57 g/mol
Boiling Point 51 °C (1013 hPa)
Vapor Pressure 350 hPa (20 °C)
Flash Point -27 °C
Refractive Index 1.3848 (20 °C, 589 nm)
Explosion Limit 1.8 - 10.1 %(V)
Density 0.84 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR