Benzylurea, Sigma-Aldrich®

Leverandør: Merck

Synonymer: N-Benzylurea

M-Clarity™ quality level = MQ200

8.01795.0100
8.01795.0100EA 1250 NOK
8.01795.0100
Benzylurea, Sigma-Aldrich®
Benzylurea
Formel: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL nummer: MFCD00007951
CAS-nummer: 538-32-9
EINECS: 208-689-1

Bestill nå

Spesifikasjon Testresultater

Solubility 17 g/l (45 °C)
Melting Point 143 - 146 °C
Molar Mass 150.17 g/mol
Hill Formula C8H10N2O
Chemical Formula C6H5CH2NHCONH2

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR