Chromium(II) chloride for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Chromium dichloride

M-Clarity™ quality level = MQ200

8.02482.0010
8.02482.0010EA 2675 NOK
8.02482.0010
Chromium(II) chloride for synthesis, Sigma-Aldrich®
Chromium(II) chloride
Formel: Cl₂Cr
Kokepunkt: 1302 °C (1013 hPa)
Smeltepunkt: 823…825 °C
Tetthet: 2,87 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010947
CAS-nummer: 10049-05-5
EINECS: 233-163-3
FN: 3260
ADR: 8,III
Merck Indeks: 13,02264

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 823 - 825 °C
Molar Mass 122.9 g/mol
Boiling Point 1300 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR