5-Chloro-o-toluidine zS, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 5-Chloro-2-methylaniline, 2-Amino-4-chlorotoluene

MQ200

8.02641.0250
8.02641.0250EA 2125 NOK
8.02641.0250
5-Chloro-o-toluidine zS, Sigma-Aldrich®
5-Chloro-o-toluidine
Formel: C₇H₈ClN
MW: 141.6 g/mol
Kokepunkt: 237
Smeltepunkt: 22
Tetthet: 1.17 g/cm³ (20 °C)
Flash punkt: 160
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007779
CAS-nummer: 95-79-4
EINECS: 202-452-6
FN: 3429
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 22 °C
Molar Mass 141.59 g/mol
Boiling Point 237 °C (1013 hPa)
Vapor Pressure 0.45 hPa (50 °C)
Flash Point 160 °C
Refractive Index 1.584 (20 °C, 589 nm)
Density 1.17 g/cm3 (20 °C)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR