Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

M-Clarity™ quality level = MQ200

8.03206.0250 8.03206.1000
8.03206.0250EA 478 NOK
8.03206.0250 8.03206.1000
Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®
Benzyl(dimethyl)amine
Formel: C₆H₅CH₂N(CH₃)₂
Kokepunkt: 183 °C (1013 hPa)
Smeltepunkt: –75 °C
Tetthet: 0,915 g/cm³ (20 °C)
Flash punkt: 53 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008329
CAS-nummer: 103-83-3
EINECS: 203-149-1
FN: 2619
ADR: 8 (3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 8 g/l (20 °C)
Melting Point -75 °C
Molar Mass 135.21 g/mol
Boiling Point 180 - 183 °C (1013 hPa)
Vapor Pressure 2.4 hPa (20 °C)
Flash Point 55 °C
Refractive Index 1.5011 (20 °C, 589 nm)
Explosion Limit 0.9 - 6.3 %(V)
Density 0.90 g/cm3 (20 °C)
pH 10 (10 g/l, H2O, 20 °C) (saturated solution)
Ignition Point 410 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR