1,4-Diklorbutan for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

8.03227.0250 8.03227.1000
8.03227.0250EA 518 NOK
8.03227.0250 8.03227.1000
1,4-Diklorbutan for synthesis, Sigma-Aldrich®
1,4-Diklorbutan
Formel: ClH₂C(CH₂)₂CH₂Cl
Kokepunkt: 153,9 °C (1013 hPa)
Smeltepunkt: –38 °C
Tetthet: 1,1457 g/cm³ (20 °C)
Flash punkt: 53 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001011
CAS-nummer: 110-56-5
EINECS: 203-778-1
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Solubility 2.4 g/l (20 °C)
Melting Point -38 °C
Molar Mass 127.01 g/mol
Boiling Point 155 °C (1013 hPa)
Vapor Pressure 5 hPa (20 °C)
Flash Point 53 °C
Refractive Index 1.4545 (20 °C, 589 nm)
Explosion Limit 1.8 - 8.9 %(V)
Density 1.14 g/cm3 (20 °C)
pH 7.3 (H2O, 20 °C)
Ignition Point 220 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR