N,N-Dimetylacetamid for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.03235.1000 8.03235.2500 8.03235.9026
8.03235.1000EA 581 NOK
8.03235.1000 8.03235.2500 8.03235.9026
N,N-Dimetylacetamid for syntese, Sigma-Aldrich®
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
Kokepunkt: 166,1 °C (1013 hPa)
Smeltepunkt: –20 °C
Tetthet: 0,937 g/cm³ (20 °C)
Flash punkt: 70 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
FN: 2810
ADR: 6.1,III
Merck Indeks: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -20 °C
Molar Mass 87.12 g/mol
Boiling Point 165 - 166 °C (1013 hPa)
Vapor Pressure 1.76 hPa (20 °C)
Flash Point 64 °C
Refractive Index 1.423 (20 °C, 589 nm)
Explosion Limit 1.7 - 11.5 %(V)
Density 0.94 g/cm3 (20 °C)
pH 4 (200 g/l, H2O, 20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR