1,2-Diklorbenzen for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: o-Dichlorobenzene

M-Clarity™ quality level = MQ200

8.03238.0100 8.03238.1000 8.03238.2500
8.03238.0100EA 229 NOK
8.03238.0100 8.03238.1000 8.03238.2500
1,2-Diklorbenzen for synthesis, Sigma-Aldrich®
1,2-Diklorbenzen
Formel: C₆H₄Cl₂
Kokepunkt: 180,5 °C (1013 hPa)
Smeltepunkt: –17 °C
Tetthet: 1,309 g/cm³ (20 °C)
Flash punkt: 66 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000535
CAS-nummer: 95-50-1
EINECS: 202-425-9
FN: 1591
ADR: 6.1,III
Merck Indeks: 13,03081

Bestill nå

Spesifikasjon Testresultater

Solubility 0.13 g/l (20 °C)
Melting Point -17 °C
Molar Mass 147 g/mol
Boiling Point 180 °C (1013 hPa)
Vapor Pressure 1.33 hPa (20 °C)
Flash Point 66 °C
Explosion Limit 2.2 - 12 %(V)
Density 1.31 g/cm3 (20 °C)
Ignition Point 640 °C
Synonyms o-Chlorobenzene
Hill Formula C6H4Cl2
Chemical Formula 1,2-(Cl)2C6H4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR