Dipropyleneglycol (mixture of isomers) for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Dipropylene glycol

M-Clarity™ quality level = MQ200

8.03265.1000
8.03265.1000EA 556 NOK
8.03265.1000
Dipropyleneglycol (mixture of isomers) for synthesis, Sigma-Aldrich®
Dipropyleneglycol (mixture of isomers)
Formel: C₆H₁₄O₃
MDL nummer: MFCD00051023
CAS-nummer: 25265-71-8
EINECS: 246-770-3

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -32 °C
Molar Mass 134.17 g/mol
Boiling Point 231 - 235 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 121 °C
Refractive Index 1.441 (20 °C, 589 nm)
Explosion Limit 2.9 - 12.6 %(V)
Density 1.02 g/cm3 (20 °C)
pH 6 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 310 °C DIN 51794

SPESIFIKASJONER

Environmentally Preferable
Miljøsertifikat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR