(±)-Epiklorhydrin zS, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane

MQ200

8.03296.8000
8.03296.8000EA 3925 NOK
8.03296.8000
(±)-Epiklorhydrin zS, Sigma-Aldrich®
(±)-Epiklorhydrin
Formel: C₃H₅ClO
MW: 92.52 g/mol
Kokepunkt: 117.9
Smeltepunkt: –57.2 °C
Tetthet: 0.9985 g/cm³ (20 °C)
Flash punkt: 28
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005132
CAS-nummer: 106-89-8
EINECS: 203-439-8
FN: 2023
ADR: 6.1 (3) II
Merck Indeks: 13,03642

Bestill nå

Spesifikasjon Testresultater

Solubility 60 g/l (20 °C)
Melting Point -57.2 °C
Molar Mass 92.52 g/mol
Boiling Point 116.5 °C (1013 hPa)
Vapor Pressure 16.5 hPa (20 °C)
Flash Point 28 °C
Refractive Index 1.4375 (20 °C, 589 nm)
Explosion Limit 2.3 - 34.4 %(V)
Density 1.18 g/cm3 (20 °C)
Ignition Point 416 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR