1,3-Diethyl-2-thiourea, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: N,N'-Diethylthiourea

M-Clarity™ quality level = MQ200

8.03520.0250 8.03520.5000 8.03520.0050
8.03520.0250EA 399 NOK
8.03520.0250 8.03520.5000 8.03520.0050
1,3-Diethyl-2-thiourea, Sigma-Aldrich®
1,3-Diethyl-2-thiourea
Formel: C₅H₁₂N₂S
Kokepunkt: 169 °C (1013 hPa)
Smeltepunkt: 75…78 °C
Tetthet: 1,118 g/cm³ (20 °C)
Flash punkt: 186 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004925
CAS-nummer: 105-55-5
EINECS: 203-308-5

Bestill nå

Spesifikasjon Testresultater

Solubility 38 g/l (20 °C)
Melting Point 75 - 78 °C
Molar Mass 132.22 g/mol
Bulk Density 735 kg/m3
Flash Point 186 °C
Density 1.12 g/cm3 (20 °C)
pH 7 (38 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR