1,3-Diphenyl-2-thiourea, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Thiocarbanilide, N,N'-Diphenylthiourea, N,N’-Diphenylthiourea

M-Clarity™ quality level = MQ200

8.03581.0100 8.03581.0500
8.03581.0100EA 363 NOK
8.03581.0100 8.03581.0500
1,3-Diphenyl-2-thiourea, Sigma-Aldrich®
1,3-Diphenyl-2-thiourea
Formel: C₁₃H₁₂N₂S
Kokepunkt: 365 °C (1013 hPa)
Smeltepunkt: 150…153 °C
Tetthet: 1,3205 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004921
CAS-nummer: 102-08-9
EINECS: 203-004-2
FN: 3077
ADR: 9,III
Merck Indeks: 13,03366

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 150 - 153 °C
Molar Mass 228.31 g/mol
Bulk Density 400 kg/m3
Density 1.30 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR