1-Heksanol ≥98% for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

M-Clarity™ quality level = MQ200

8.04393.0100 8.04393.1000 8.04393.2500 8.04393.9020
8.04393.0100EA 306 NOK
8.04393.0100 8.04393.1000 8.04393.2500 8.04393.9020
1-Heksanol ≥98% for synthesis, Sigma-Aldrich®
1-Heksanol
Formel: H₃C(CH₂)₄CH₂OH
Kokepunkt: 156 °C (1013 hPa)
Smeltepunkt: –45 °C
Tetthet: 0,8322 g/cm³ (20 °C)
Flash punkt: 60 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002982
CAS-nummer: 111-27-3
EINECS: 203-852-3
FN: 2282
ADR: 3,III
Merck Indeks: 13,04715
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC, area%) ≥ 98 %
Density (d 20 °C/ 4 °C) 0.818 - 0.819
Identity (IR) passes test
Synonyms 1-Hexanol
Chemical Formula CH3(CH2)5OH

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR