m-Anisaldehyd for synthesis, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 3-Methoxybenzaldehyde

M-Clarity™ quality level = MQ200

8.05796.9020
8.05796.9020EA 0 NOK
8.05796.9020
m-Anisaldehyd for synthesis, Sigma-Aldrich®
m-Anisaldehyd
Formel: C₈H₈O₂
MW: 136.15 g/mol
Kokepunkt: 228…229 °C (1013 hPa)
Tetthet: 1.1149 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003361
CAS-nummer: 591-31-1
EINECS: 209-712-8

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Molar Mass 136.15 g/mol
Boiling Point 230 - 233 °C (1013 hPa)
Vapor Pressure 1.3 hPa (73 °C)
Flash Point 110 °C
Refractive Index 1.5523 (20 °C, 589 nm)
Density 1.12 g/cm3 (20 °C)
Ignition Point 225 °C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR