1-Metylimidazol ≥99.0% (by GC) for syntese

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.05852.0100 8.05852.1000 8.05852.0025
8.05852.0100EA 466 NOK
8.05852.0100 8.05852.1000 8.05852.0025
1-Metylimidazol ≥99.0% (by GC) for syntese
1-Metylimidazol
Formel: C₄H₆N₂
Kokepunkt: 198 °C (1013 hPa)
Smeltepunkt: –2 °C
Tetthet: 1,03 g/cm³ (20 °C)
Flash punkt: 92 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005292
CAS-nummer: 616-47-7
EINECS: 210-484-7
FN: 2922
ADR: 8 (6.1),II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -2 °C
Molar Mass 82.10 g/mol
Boiling Point 198 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 92 °C
Explosion Limit 2.7 - 15.7 %(V)
Density 1.035 g/cm3 (20 °C)
pH 9.5 - 10.5 (50 g/l, H2O, 20 °C)
Ignition Point 525 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR