2-Metyl-2-butanol, EMPLURA®, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.06193.1000
8.06193.1000EA 756 NOK
8.06193.1000
2-Metyl-2-butanol, EMPLURA®, Supelco®
2-Metyl-2-butanol
Formel: C₅H₁₂O
Kokepunkt: 102 °C (1013 hPa)
Tetthet: 0,806 g/cm³ (20 °C)
Flash punkt: 20 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004478
CAS-nummer: 75-85-4
EINECS: 200-908-9
FN: 1105
ADR: 3,II
Merck Indeks: 13,07218

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR