Natriumtetrahydroborat, finkornet for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.06373.0025 8.06373.0100 8.06373.0500 8.06373.0010
8.06373.0025EA 554 NOK
8.06373.0025 8.06373.0100 8.06373.0500 8.06373.0010
Natriumtetrahydroborat, finkornet for syntese, Sigma-Aldrich®
Sodium borohydride
Formel: NaBH₄
Smeltepunkt: 400 °C
Tetthet: 1,07 g/cm³ (20 °C)
Flash punkt: 69 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2
EINECS: 241-004-4
FN: 1426
ADR: 4.3,I
Merck Indeks: 13,08664

Bestill nå

Spesifikasjon Testresultater

Solubility 550 g/l (25 °C) (slow decomposition)
Melting Point 400 °C (slow decomposition)
Molar Mass 37.83 g/mol
Bulk Density 350 - 500 kg/m3
Boiling Point 500 °C (decomposition)
Explosion Limit 3.02 %(V)
Density 1.07 g/cm3 (20 °C)
pH 11 (10 g/l, H2O, 20 °C)
Ignition Point 220 °C
Hazard Class 4.3
Hazard Class Description Dangerous when wet material

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR