(S)-(-)-α-Methylbenzylamine for racemat separation, for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

M-Clarity™ quality level = MQ200

8.07047.0025 8.07047.0250 8.07047.0010
8.07047.0025EA 757 NOK
8.07047.0025 8.07047.0250 8.07047.0010
(S)-(-)-α-Methylbenzylamine for racemat separation, for synthesis, Sigma-Aldrich®
(S)-(-)-α-Methylbenzylamine
Formel: C₈H₁₁N
MW: 121.18 g/mol
Kokepunkt: 187
Smeltepunkt: –10 °C
Tetthet: 0.95 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064406
CAS-nummer: 2627-86-3
EINECS: 220-098-0
FN: 2922
ADR: 8 (6.1) II
Merck Indeks: 12,06106

Bestill nå

Spesifikasjon Testresultater

Solubility 42 g/l (20 °C)
Melting Point -10 °C
Molar Mass 121.18 g/mol
Boiling Point 187 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 71 °C
Refractive Index 1.5259 (20 °C, 589 nm)
Density 0.95 g/cm3 (20 °C)
Ignition Point 355 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR