Palladium (II) oksid hydrogenation catalyst, for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.07107.0001 8.07107.8500
8.07107.0001EA 4270 NOK
8.07107.0001 8.07107.8500
Palladium (II) oksid hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Palladium (II) oksid
Formel: OPd
Smeltepunkt: 870 °C
Tetthet: 8,73 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011172
CAS-nummer: 1314-08-5
EINECS: 215-218-3
FN: 1479
ADR: 5.1,II
Merck Indeks: 12,07125

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Molar Mass 122.42 g/mol
Hill Formula OPd
Chemical Formula PdO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR