Palladium(II)klorid, vannfri for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.07110.0001 8.07110.0005 8.07110.0010 8.07110.0050 8.07110.8500
8.07110.0001EA 2750 NOK
8.07110.0001 8.07110.0005 8.07110.0010 8.07110.0050 8.07110.8500
Palladium(II)klorid, vannfri for syntese, Sigma-Aldrich®
Palladium(II)klorid
Formel: PdCl₂
Smeltepunkt: 678 °C
Tetthet: 4 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003558
CAS-nummer: 7647-10-1
EINECS: 231-596-2
FN: 3260
ADR: 8,II
Merck Indeks: 13,07058

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 680 °C
Molar Mass 177.33 g/mol
Density 4.0 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR