Pelargonaldehyd for syntese, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 1-Nonanal, Nonylaldehyde, Nonanal, Nonaldehyde

M-Clarity™ quality level = MQ200

8.07166.0050
8.07166.0050EA 478 NOK
8.07166.0050
Pelargonaldehyd for syntese, Sigma-Aldrich®
Pelargonaldehyd
Formel: C₉H₁₈O
MW: 142,24 g/mol
Kokepunkt: 190 °C (1013 hPa)
Smeltepunkt: –40 °C
Tetthet: 0,83 g/cm³ (20 °C)
Flash punkt: 76 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007030
CAS-nummer: 124-19-6
EINECS: 204-688-5

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Molar Mass 142.24 g/mol
Boiling Point 93 °C (31 hPa)
Flash Point 76 °C
Refractive Index 1.424 (20 °C, 589 nm)
Density 0.83 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR