Platinium(IV) oxide hydrate hydrogenation catalyst, for syntese, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

8.07346.0001 8.07346.8500
8.07346.0001EA 2910 NOK
8.07346.0001 8.07346.8500
Platinium(IV) oxide hydrate hydrogenation catalyst, for syntese, Sigma-Aldrich®
Platinium(IV) oxide hydrate
Formel: O₂Pt·H₂O
Smeltepunkt: 450 °C
Tetthet: 10,2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066964
CAS-nummer: 52785-06-5
EINECS: 215-223-0
FN: 3085
ADR: 5.1 (8),II
Merck Indeks: 12,07686

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 450 °C
Molar Mass 227.08 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR