1-Pentanol for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol

M-Clarity™ quality level = MQ200

8.07500.0100 8.07500.1000 8.07500.2500 8.07500.9160
8.07500.0100EA 145 NOK
8.07500.0100 8.07500.1000 8.07500.2500 8.07500.9160
1-Pentanol for synthesis, Sigma-Aldrich®
1-Pentanol
Formel: H₃C(CH₂)₃CH₂OH
Kokepunkt: 137,9 °C (1013 hPa)
Smeltepunkt: –79 °C
Tetthet: 0,81 g/cm³ (20 °C)
Flash punkt: 48 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002977
CAS-nummer: 71-41-0
EINECS: 200-752-1
FN: 1105
ADR: 3,III
Merck Indeks: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 22 g/l (20 °C)
Melting Point -79 °C
Molar Mass 88.15 g/mol
Boiling Point 138 °C (1013 hPa)
Vapor Pressure 2.6 hPa (20 °C)
Flash Point 49 °C
Refractive Index 1.41 (20 °C, 589 nm)
Explosion Limit 1.6 %(V)
Density 0.815 g/cm3 (20 °C)
pH (22 g/l, H2O, 20 °C) neutral
Ignition Point 300 °C DIN 51794
Evaporation Number 2.7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR