1-Propanethiol for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane

M-Clarity™ quality level = MQ200

8.07529.0005 8.07529.0250
8.07529.0005EA 375 NOK
8.07529.0005 8.07529.0250
1-Propanethiol for synthesis, Sigma-Aldrich®
1-Propanethiol
Formel: C₃H₈S
Kokepunkt: 67 °C (1013 hPa)
Smeltepunkt: –113 °C
Tetthet: 0,82995 g/cm³ (20 °C)
Flash punkt: –14 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004900
CAS-nummer: 107-03-9
EINECS: 203-455-5
FN: 2402
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Solubility 1.9 g/l (25 °C)
Melting Point -113 °C
Molar Mass 76.16 g/mol
Boiling Point 67 - 68 °C (1013 hPa)
Vapor Pressure 165 hPa (20 °C)
Flash Point -34 °C
Refractive Index 1.438 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR