Antimony(III) fluoride zS, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Antimony trifluoride

MQ200

8.07839.0005
8.07839.0005EA 307 NOK
8.07839.0005
Antimony(III) fluoride zS, Sigma-Aldrich®
Antimony(III) fluoride
Formel: F₃Sb
MW: 178.76 g/mol
Kokepunkt: 376
Smeltepunkt: 292
Tetthet: 4.38 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011218
CAS-nummer: 7783-56-4
EINECS: 232-009-2
FN: 1549
ADR: 6.1 III
Merck Indeks: 13,00715

Bestill nå

Spesifikasjon Testresultater

Solubility 4430 g/l (20 °C) Hydrolysis
Melting Point 292 °C
Molar Mass 178.74 g/mol
Bulk Density 1500 kg/m3
Boiling Point 376 °C (1013 hPa)
Density 4.38 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR