Sulfolan ≥99.0% for synthesis, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: Tetrahydrotiofen-1,1-dioksid

M-Clarity™ quality level = MQ200

8.07993.0250 8.07993.1000
8.07993.0250EA 549 NOK
8.07993.0250 8.07993.1000
Sulfolan ≥99.0% for synthesis, Sigma-Aldrich®
Sulfolan
Formel: C₄H₈O₂S
Kokepunkt: 285 °C (1013 hPa)
Smeltepunkt: 26 °C
Tetthet: 1,26 g/cm3 (30 °C)
Flash punkt: 177 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005484
CAS-nummer: 126-33-0
EINECS: 204-783-1
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Assay (GC, area%) ≥ 99.0 % (a/a)
Water (K. F.) ≤ 0.20 %
Identity (IR) passes test
Boiling Point 285 °C (1013 hPa)
Vapor Pressure 0.035 hPa (30 °C)
Flash Point 177 °C
Refractive Index 1.484 (20 °C, 589 nm)
Density 1.26 g/cm3 (30 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR