4,4'-Bis(dimetylamino)bensofenon for synthesis, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

M-Clarity™ quality level = MQ200

8.08125.0250 8.08125.1000
8.08125.0250EA 797 NOK
8.08125.0250 8.08125.1000
4,4'-Bis(dimetylamino)bensofenon for synthesis, Sigma-Aldrich®
4,4'-Bis(dimetylamino)bensofenon
Formel: C₁₇H₂₀N₂O
Kokepunkt: 428 °C (1013 hPa)
Smeltepunkt: 172 °C
Flash punkt: 220 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008312
CAS-nummer: 90-94-8
EINECS: 202-027-5
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,06265

Bestill nå

Spesifikasjon Testresultater

Solubility 0.4 g/l (20 °C)
Melting Point 172 - 175 °C
Molar Mass 268.35 g/mol
Bulk Density 400 kg/m3
Boiling Point 360 °C (1013 hPa)
Flash Point 220 °C
Ignition Point 480 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR