3,3'-Thiodipropionic acid, Sigma-Aldrich®

Leverandør: Merck

Synonymer: 3,3-Thiodipropionic acid

M-Clarity™ quality level = MQ200

8.08626.0250
8.08626.0250EA 523 NOK
8.08626.0250
3,3'-Thiodipropionic acid, Sigma-Aldrich®
3,3'-Thiodipropionic acid
Formel: C₆H₁₀O₄S
MW: 178.21 g/mol
Kokepunkt: 347
Smeltepunkt: 128…131 °C
Tetthet: 1.496 g/cm³ (20 °C)
MDL nummer: MFCD00002781
CAS-nummer: 111-17-1
EINECS: 203-841-3
Merck Indeks: 12,09468

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR